Gene Transcripts

Gene Symbol
Igtp
Entrez Gene ID
16145
Gene Name
interferon gamma induced GTPase
Gene Aliases
AW558444, Irgm3
Chromosome Location
Chr. 11: 58199556 - 58207592 on Build GRCm38
UniGene ID
Mm.33902
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018738.4 NP_061208.3 3 1105
GenBank mRNA AK002843.1 - 3 1043
AK088315.1 - 3 1104
AK149817.1 - 3 1034
AK151779.1 - 3 1045
AK151860.1 - 3 1026
AK152337.1 - 4 877
AK152483.1 - 3 1026
AK153436.1 - 3 1045
AK165065.1 - 4 932
AK169307.1 - 3 1064
AY225483.1 - 2 670
BC132557.1 - 2 733 40130834
BC138607.1 - 2 733 8861627
BC145346.1 - 3 534
U53219.1 - 3 1017

View Full Product Details