Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018788.3 | NP_061258.2 | 7 | 3573 | |
GenBank mRNA | AF083550.1 | - | 5 | 3363 | |
AK137842.1 | - | 5 | 2650 | ||
AK190300.1 | - | 1 | 191 | ||
BC058526.1 | - | 5 | 2964 | ||
BC065073.1 | - | 5 | 3038 | 6834580 |