Gene Transcripts
- Gene Symbol
- Wwox
- Entrez Gene ID
- 80707
- Gene Name
- WW domain-containing oxidoreductase
- Gene Aliases
- 5330426P09Rik, 9030416C10Rik, WOX1
- Chromosome Location
- Chr. 8: 114439630 - 115352712 on Build GRCm38
- UniGene ID
- Mm.440420
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019573.3 |
NP_062519.2
|
2, 3 |
297 |
|
GenBank mRNA |
AF187014.1 |
- |
2, 3 |
248 |
|
AK018507.1 |
- |
2, 3 |
227 |
|
AK019911.1 |
- |
2, 3 |
255 |
|
AK046903.1 |
- |
2, 3 |
247 |
|
AK050541.1 |
- |
2, 3 |
248 |
|
AK078528.1 |
- |
2, 3 |
250 |
|
AK083096.1 |
- |
2, 3 |
229 |
|
BC014716.1 |
- |
2, 3 |
253 |
4241066 |
CJ058964.1 |
- |
2, 3 |
298 |
|
View Full Product Details