Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282428.1 | NP_001269357.1 | 3 | 1346 | |
NM_002252.4 | NP_002243.3 | 3 | 1296 | ||
XM_011532825.1 | XP_011531127.1 | 4 | 1657 | ||
XM_017004059.1 | XP_016859548.1 | 3 | 1246 | ||
GenBank mRNA | AF043472.1 | - | 2 | 1026 | |
AK075088.1 | - | 1 | 654 | ||
AK225833.1 | - | 3 | 1346 | ||
AK314451.1 | - | 3 | 1296 | ||
BC004148.2 | - | 3 | 1233 | 2958834 | |
BC004987.1 | - | 3 | 1233 | 2957877 | |
BC015947.2 | - | 3 | 1285 | 3919594 |