Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243748.1 | NP_001230677.1 | 3 | 470 | |
NM_019713.4 | NP_062687.1 | 2 | 414 | ||
GenBank mRNA | AF132851.1 | - | 2 | 247 | |
AK146736.1 | - | 2 | 213 | ||
BC002173.1 | - | 2 | 341 | 3486727 | |
BC052760.1 | - | 1 | 784 | ||
CD564987.1 | - | 3 | 285 | ||
CX224234.1 | - | 2 | 366 | ||
EU314810.1 | - | 3 | 428 |