Gene Transcripts

Gene Symbol
Ruvbl1
Entrez Gene ID
56505
Gene Name
RuvB-like protein 1
Gene Aliases
2510009G06Rik, Pontin52, Tip49a
Chromosome Location
Chr. 6: 88465423 - 88497566 on Build GRCm38
UniGene ID
Mm.42195
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019685.2 NP_062659.1 3 443
GenBank mRNA AF100694.1 - 3 482
AK012170.1 - 3 391
AK132783.1 - 3 425
AK145062.1 - 392
AK146258.1 - 3 435
AK146374.1 - 3 436
AK156078.1 - 3 413
AK198661.1 - 2 133
BC004718.1 - 3 409 3582426

View Full Product Details