Gene Transcripts

Gene Symbol
Cpxm1
Entrez Gene ID
56264
Gene Name
carboxypeptidase X 1 (M14 family)
Gene Aliases
AA986902, Cpx-1, Cpx1, Cpxm
Chromosome Location
Chr. 2: 130390775 - 130397629 on Build GRCm38
UniGene ID
Mm.112701
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019696.2 NP_062670.2 3 472
GenBank mRNA AF077738.1 - 3 417
BC003713.1 - 3 405 3593211

View Full Product Details