Gene Transcripts

Gene Symbol
Tex21
Entrez Gene ID
80384
Gene Name
testis expressed gene 21
Gene Aliases
4931406F04Rik, 4931412D23Rik, 4931421K24Rik, tsec-2
Chromosome Location
Chr. 12: 76198687 - 76246758 on Build GRCm38
UniGene ID
Mm.78551
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159532.1 NP_001153004.1 2 411
NM_019784.5 NP_062758.3 2 411
GenBank mRNA AB000682.1 - 2 408
AK016434.1 - 2 443
AK016448.1 - 2 401
AK016471.1 - 2 382
BC100432.1 - 2 411 30917866

View Full Product Details