Gene Transcripts

Gene Symbol
Snd1
Entrez Gene ID
56463
Gene Name
staphylococcal nuclease and tudor domain containing 1
Gene Aliases
AL033314, Tudor-SN
Chromosome Location
Chr. 6: 28480315 - 28935162 on Build GRCm38
UniGene ID
Mm.439987
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019776.2 NP_062750.2 4, 5 637
GenBank mRNA AB021491.1 - 4, 5 594
AK088028.1 - 4, 5 596
AK133837.1 - 4, 5 643
AK159837.1 - 4, 5 602
AK161591.1 - 4, 5 591
AK162432.1 - 4, 5 578
AK167576.1 - 4, 5 627
BC007126.1 - 4, 5 584 3592548
BM944959.1 - 3, 4 315

View Full Product Details