Gene Transcripts
- Gene Symbol
- Stat2
- Entrez Gene ID
- 20847
- Gene Name
- signal transducer and activator of transcription 2
- Gene Aliases
- 1600010G07Rik, AW496480
- Chromosome Location
- Chr. 10: 128270559 - 128292852 on Build GRCm38
- UniGene ID
- Mm.293120
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019963.1 |
NP_064347.1
|
23, 24 |
2778 |
|
GenBank mRNA |
AF088862.1 |
- |
23, 24 |
2651 |
|
AF187231.1 |
- |
23, 24 |
2775 |
|
AF206162.1 |
- |
23, 24 |
2778 |
|
AK005415.1 |
- |
1, 2 |
133 |
|
AK030903.1 |
- |
16, 17 |
1870 |
|
AK149924.1 |
- |
23, 24 |
2818 |
|
AK150070.1 |
- |
23, 24 |
2790 |
|
AK153405.1 |
- |
21, 22 |
2425 |
|
BC064827.1 |
- |
13, 14 |
1655 |
|
View Full Product Details