Gene Transcripts

Gene Symbol
Psg23
Entrez Gene ID
56868
Gene Name
pregnancy-specific glycoprotein 23
Gene Aliases
1620401C02Rik, cea11
Chromosome Location
Chr. 7: 18606343 - 18616501 on Build GRCm38
UniGene ID
Mm.333617
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020261.4 NP_064657.2 5 1854
GenBank mRNA AK018828.1 - 5 1855

View Full Product Details