Gene Transcripts

Gene Symbol
AQP9
Entrez Gene ID
366
Gene Name
aquaporin 9
Gene Aliases
AQP-9, HsT17287, SSC1, T17287
Chromosome Location
Chr. 15: 58138169 - 58185911 on Build GRCh38
UniGene ID
Hs.104624
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320635.1 NP_001307564.1 5 935
NM_001320636.1 NP_001307565.1 6 785
NM_020980.4 NP_066190.2 6 1153
GenBank mRNA AB008775.1 - 6 999
AF016495.1 - 6 1011
AK300055.1 - 6 758
AK313214.1 - 6 1078
BC026258.1 - 6 968 4734559
CR542027.1 - 6 783

View Full Product Details