Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020590.4 | NP_065615.1 | 4 | 563 | |
GenBank mRNA | AB041648.1 | - | 4 | 587 | |
AF180518.1 | - | 4 | 590 | ||
AK006103.1 | - | 4 | 362 | ||
AK011975.1 | - | 4 | 558 | ||
AK014083.1 | - | 4 | 377 | ||
AK015049.1 | - | 4 | 380 | ||
AK018687.1 | - | 4 | 564 | ||
AK159015.1 | - | 4 | 558 | ||
AK167500.1 | - | 4 | 556 | ||
AK168921.1 | - | 4 | 595 | ||
BC004602.1 | - | 4 | 498 | 3585132 | |
BC024706.1 | - | 4 | 562 | 3711973 |