Gene Transcripts

Gene Symbol
Klrc3
Entrez Gene ID
58179
Gene Name
killer cell lectin-like receptor subfamily C, member 3
Gene Aliases
Klrc2, Nkg2e
Chromosome Location
Chr. 6: 129636263 - 129643511 on Build GRCm38
UniGene ID
Mm.358684
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021378.1 NP_067353.1 3 303
GenBank mRNA AF195779.1 - 3 303

View Full Product Details