Gene Transcripts

Gene Symbol
Nap1l5
Entrez Gene ID
58243
Gene Name
nucleosome assembly protein 1-like 5
Gene Aliases
1110020M21Rik
Chromosome Location
Chr. 6: 58905233 - 58907126 on Build GRCm38
UniGene ID
Mm.389747
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021432.2 NP_067407.1 1 428
GenBank mRNA AB041556.1 - 1 367
AK003875.1 - 1 134
BC046332.1 - 1 378 6491916
BU526293.1 - 1 431

View Full Product Details