Gene Transcripts
- Gene Symbol
- KCNIP1
- Entrez Gene ID
- 30820
- Gene Name
- potassium voltage-gated channel interacting protein 1
- Gene Aliases
- KCHIP1, VABP
- Chromosome Location
- Chr. 5: 170353055 - 170736632 on Build GRCh38
- UniGene ID
- Hs.484111
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001034837.2 |
NP_001030009.1
|
3, 4 |
753 |
|
NM_001034838.2 |
NP_001030010.1
|
2, 3 |
600 |
|
NM_001278339.1 |
NP_001265268.1
|
2, 3 |
720 |
|
NM_001278340.1 |
NP_001265269.1
|
3, 4 |
882 |
|
NM_014592.3 |
NP_055407.1
|
2, 3 |
720 |
|
XM_017009407.1 |
XP_016864896.1
|
3, 4 |
724 |
|
GenBank mRNA |
AF199597.1 |
- |
2, 3 |
183 |
|
AI884645.1 |
- |
2, 3 |
486 |
|
AK074390.1 |
- |
2, 3 |
316 |
|
AK301775.1 |
- |
2, 3 |
375 |
|
AY170821.1 |
- |
2, 3 |
396 |
|
AY780424.1 |
- |
3, 4 |
558 |
|
BC035032.1 |
- |
2, 3 |
725 |
|
BC050375.1 |
- |
2, 3 |
487 |
5752207 |
DQ148476.1 |
- |
2, 3 |
210 |
|
DQ148477.1 |
- |
2, 3 |
183 |
|
DQ148478.1 |
- |
3, 4 |
216 |
|
DQ148479.1 |
- |
2, 3 |
183 |
|
View Full Product Details