Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163359.1 | NP_001156831.1 | 3 | 816 | |
NM_001163360.1 | NP_001156832.1 | 2 | 686 | ||
NM_021891.3 | NP_068691.2 | 3 | 806 | ||
GenBank mRNA | AF263914.1 | - | 2 | 720 | |
AK051324.1 | - | 3 | 914 | ||
AK051874.1 | - | 3 | 776 | ||
AK086769.1 | - | 3 | 806 | ||
AK088574.1 | - | 3 | 789 | ||
AK143850.1 | - | 3 | 785 | ||
AK148994.1 | - | 1 | 839 | ||
BC051942.1 | - | 3 | 759 | 6401467 | |
BC052415.1 | - | 3 | 759 | 6401467 |