Gene Transcripts

Gene Symbol
Nisch
Entrez Gene ID
64652
Gene Name
nischarin
Gene Aliases
1200007D05Rik, 3202002H23Rik, AW494485, I-1
Chromosome Location
Chr. 14: 31170928 - 31206826 on Build GRCm38
UniGene ID
Mm.298728
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022656.2 NP_073147.2 10 1268
GenBank mRNA AF315344.1 - 6 703
AK004628.1 - 10 1105
AK036043.1 - 10 1123
AK049791.1 - 10 1123
AK086880.1 - 9, 10 1020
AK160990.1 - 10 1142
BC003270.1 - 10 1130
BC066838.1 - 10 1126
BC089329.1 - 5 487
BC108364.1 - 5 463
BC144980.1 - 6 597

View Full Product Details