Gene Transcripts
- Gene Symbol
- Raly
- Entrez Gene ID
- 19383
- Gene Name
- hnRNP-associated with lethal yellow
- Gene Aliases
- AI663842, Merc
- Chromosome Location
- Chr. 2: 154791097 - 154871010 on Build GRCm38
- UniGene ID
- Mm.221440
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001139511.1 |
NP_001132983.1
|
3, 4 |
785 |
|
NM_001139512.1 |
NP_001132984.1
|
4, 5 |
551 |
|
NM_001139513.1 |
NP_001132985.1
|
4, 5 |
833 |
|
NM_023130.3 |
NP_075619.2
|
4, 5 |
868 |
|
GenBank mRNA |
AF148458.1 |
- |
2, 3 |
88 |
|
AK014356.1 |
- |
4, 5 |
621 |
|
AK135386.1 |
- |
3, 4 |
544 |
|
AK152290.1 |
- |
4, 5 |
627 |
|
AK154787.1 |
- |
3, 4 |
542 |
|
AK168781.1 |
- |
4, 5 |
625 |
|
BC004851.1 |
- |
3, 4 |
529 |
2810841 |
BC016587.1 |
- |
4, 5 |
617 |
2811315 |
CX214615.1 |
- |
4, 5 |
576 |
|
L17076.1 |
- |
4, 5 |
598 |
|
S72641.1 |
- |
4, 5 |
819 |
|
View Full Product Details