Gene Transcripts

Gene Symbol
CLIC5
Entrez Gene ID
53405
Gene Name
chloride intracellular channel 5
Gene Aliases
DFNB102, DFNB103, MST130, MSTP130
Chromosome Location
Chr. 6: 45898451 - 46129809 on Build GRCh38
UniGene ID
Hs.485489
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114086.1 NP_001107558.1 6 2045
NM_016929.4 NP_058625.2 6 1809
NR_045672.1 3 1387
NR_045673.1 5 1461
XM_017010953.1 XP_016866442.1 7 2290
GenBank mRNA AF176920.1 - 6 1609
AF216941.1 - 6 1759
AK075163.1 - 6 1660
AK097048.1 - 6 2045
BC035968.1 - 6 1777 4611102
DQ679794.1 - 6 1777

View Full Product Details