Gene Transcripts

Gene Symbol
Ly6g6c
Entrez Gene ID
68468
Gene Name
lymphocyte antigen 6 complex, locus G6C
Gene Aliases
1110003M04Rik, AU016360, G6c, NG24
Chromosome Location
Chr. 17: 35067325 - 35070048 on Build GRCm38
UniGene ID
Mm.215096
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023463.3 NP_075952.1 3 358
GenBank mRNA AJ315546.1 - 3 293
AJ315547.1 - 2 635
AK003375.1 - 3 359
BC116365.1 - 3 324 40100048
BC116366.1 - 3 322 40100049

Gene Symbol
Catsper3
Entrez Gene ID
76856
Gene Name
cation channel, sperm associated 3
Gene Aliases
4921522D01Rik, CACRC
Chromosome Location
Chr. 13: 55784579 - 55824942 on Build GRCm38
UniGene ID
Mm.159795
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC089518.1 - 177

View Full Product Details