Gene Transcripts

Gene Symbol
Trim12a
Entrez Gene ID
76681
Gene Name
tripartite motif-containing 12A
Gene Aliases
2310043C01Rik, Trim12
Chromosome Location
Chr. 7: 104299893 - 104315849 on Build GRCm38
UniGene ID
Mm.458309
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023835.2 NP_076324.2 2 352
GenBank mRNA AF220126.1 - 2 445
AK009769.1 - 2 314
BC094899.1 - 2 312 30041200

View Full Product Details