Gene Transcripts

Gene Symbol
Slc29a3
Entrez Gene ID
71279
Gene Name
solute carrier family 29 (nucleoside transporters), member 3
Gene Aliases
4933435C21Rik, AW987637, Ent3
Chromosome Location
Chr. 10: 60712072 - 60752850 on Build GRCm38
UniGene ID
Mm.284462
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023596.3 NP_076085.1 2, 3 341
GenBank mRNA AF326986.1 - 2, 3 352
AK034133.1 - 2, 3 337
AK157868.1 - 2, 3 342
BC137864.1 - 2, 3 324 8860883
BC144983.1 - 2, 3 324
CV557129.1 - 1, 2 211

View Full Product Details