Gene Transcripts

Gene Symbol
Snx5
Entrez Gene ID
69178
Gene Name
sorting nexin 5
Gene Aliases
0910001N05Rik, 1810032P22Rik, AU019504, D2Ertd52e
Chromosome Location
Chr. 2: 144250123 - 144270902 on Build GRCm38
UniGene ID
Mm.273379
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199188.1 NP_001186117.1 4 442
NM_024225.5 NP_077187.1 3 353
GenBank mRNA AK007676.1 - 3 301
AK029463.1 - 3 313
AK031873.1 - 4 334
AK049129.1 - 4 334
AK078776.1 - 4 335
AK144710.1 - 4 335
AK152311.1 - 4 378
AK159904.1 - 3 258
AK167362.1 - 3 273
AK169545.1 - 3 574
AK169786.1 - 4 334
AK177366.1 - 2 167
AK187575.1 - 2 165
AK214265.1 - 2 166
BC002242.1 - 4 411 3492262

View Full Product Details