Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001281726.1 | NP_001268655.1 | 2 | 276 | |
XM_006713314.3 | XP_006713377.1 | 3 | 556 | ||
XM_011534041.2 | XP_011532343.1 | 3 | 393 | ||
XM_017007088.1 | XP_016862577.1 | 4 | 669 | ||
GenBank mRNA | AK294301.1 | - | 4 | 408 | |
AK297033.1 | - | 2 | 268 | ||
AK301838.1 | - | 2 | 468 | ||
AK313252.1 | - | 2 | 259 | ||
BC012099.1 | - | 2 | 202 | 4559618 | |
BE814688.1 | - | 2 | 299 | ||
BM919775.1 | - | 2 | 269 | ||
BP326502.1 | - | 2 | 276 | ||
U03274.1 | - | 2 | 205 |