Gene Transcripts

Gene Symbol
Ier3ip1
Entrez Gene ID
66191
Gene Name
immediate early response 3 interacting protein 1
Gene Aliases
1110057H19Rik, AI644142, AL022842, AL022863, AV026606
Chromosome Location
Chr. 18: 76930027 - 76941614 on Build GRCm38
UniGene ID
Mm.28593
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025409.3 NP_079685.1 3 685
GenBank mRNA AK004281.1 - 3 669
BC021512.1 - 3 672 5351700
CF951253.1 - 2 416

View Full Product Details