Gene Transcripts

Gene Symbol
Llph
Entrez Gene ID
66225
Gene Name
LLP homolog, long-term synaptic facilitation (Aplysia)
Gene Aliases
1190005P17Rik
Chromosome Location
Chr. 10: 120227060 - 120232071 on Build GRCm38
UniGene ID
Mm.291755
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025431.2 NP_079707.1 2 150
GenBank mRNA AK004503.1 - 2 150
AK007375.1 - 2 133
AK158700.1 - 2 128
BC024098.1 - 2 151 5148362
BC096681.1 - 2 149 1332438
BC106103.1 - 136 3661552

View Full Product Details