Gene Transcripts

Gene Symbol
PRKCQ
Entrez Gene ID
5588
Gene Name
protein kinase C theta
Gene Aliases
PRKCT, nPKC-theta
Chromosome Location
Chr. 10: 6393038 - 6580646 on Build GRCh38
UniGene ID
Hs.498570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242413.2 NP_001229342.1 11 1242
NM_001282644.1 NP_001269573.1 11 1240
NM_001282645.1 NP_001269574.1 10 1181
NM_001323265.1 NP_001310194.1 11 1297
NM_001323266.1 NP_001310195.1 10 1179
NM_001323267.1 NP_001310196.1 11 1240
NM_006257.4 NP_006248.1 11 1242
XM_005252496.4 XP_005252553.1 11 1342
XM_005252497.4 XP_005252554.1 11 1344
XM_006717465.3 XP_006717528.1 13 3737
XM_017016410.1 XP_016871899.1 12 3648
XM_017016411.1 XP_016871900.1 12 3676
GenBank mRNA AK293935.1 - 10 1119
AK308099.1 - 11 1215
AK312411.1 - 11 1192
AY702977.1 - 10 1142
BC101465.1 - 10 1161 8068971
BC113359.1 - 10 1161 8322411
BX647657.1 - 10 1327
L01087.1 - 11 1236
L07032.1 - 10 1163

View Full Product Details