Gene Transcripts

Gene Symbol
Ssr2
Entrez Gene ID
66256
Gene Name
signal sequence receptor, beta
Gene Aliases
1500032E05Rik, AI315033, AU020133, TLAP, TRAPB, TRAPbeta
Chromosome Location
Chr. 3: 88579584 - 88588420 on Build GRCm38
UniGene ID
Mm.486239
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025448.3 NP_079724.1 5 1007
GenBank mRNA AK005340.1 - 6 878
AK007538.1 - 3 474
AK007720.1 - 3 474
AK010565.1 - 6 845
AK012281.1 - 5 835
AK020145.1 - 6 828
AK050505.1 - 5 1008
AK084530.1 - 6 845
AK217251.1 - 2 390
BC010214.1 - 5 991 3497894

View Full Product Details