Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164355.1 | NP_001157827.1 | 3, 4 | 454 | |
NM_025581.4 | NP_079857.3 | 3, 4 | 518 | ||
GenBank mRNA | AK006442.1 | - | 3, 4 | 415 | |
AK013233.1 | - | 3, 4 | 416 | ||
AK021226.1 | - | 3, 4 | 418 | ||
AK172500.1 | - | 3, 4 | 421 | ||
BC019940.1 | - | 3, 4 | 369 | 3709649 | |
DQ926863.1 | - | 3, 4 | 396 | ||
DQ926864.1 | - | 3, 4 | 460 |