Gene Transcripts

Gene Symbol
Mad2l1bp
Entrez Gene ID
66591
Gene Name
MAD2L1 binding protein
Gene Aliases
0610009D16Rik, 2510031P20Rik, AU018945, AW550977, C76045, CMT2A, CMT2B, Mad2lbp, mKIAA0110
Chromosome Location
Chr. 17: 46147385 - 46153551 on Build GRCm38
UniGene ID
Mm.272024
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025649.3 NP_079925.2 2 269
GenBank mRNA AK002377.1 - 2 255
AK004224.1 - 2 223
AK007974.1 - 2 221
AK156569.1 - 2 272
AK220322.1 - 2 238
BC005768.1 - 2 227 2649017

View Full Product Details