Gene Transcripts

Gene Symbol
Ccdc59
Entrez Gene ID
52713
Gene Name
coiled-coil domain containing 59
Gene Aliases
2300004H16Rik, D10Ertd718e
Chromosome Location
Chr. 10: 105841479 - 105847510 on Build GRCm38
UniGene ID
Mm.339224
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025602.3 NP_079878.1 1, 2 184
GenBank mRNA AK009028.1 - 1, 2 551
AK012917.1 - 1, 2 167
AK019970.1 - 1, 2 185
AK047009.1 - 1, 2 589
AK135550.1 - 1, 2 209
BC013721.1 - 1, 2 567
BC027401.1 - 1, 2 169 4948072

View Full Product Details