Gene Transcripts

Gene Symbol
CP
Entrez Gene ID
1356
Gene Name
ceruloplasmin (ferroxidase)
Gene Aliases
CP-2
Chromosome Location
Chr. 3: 149162410 - 149222050 on Build GRCh38
UniGene ID
Hs.558314
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000096.3 NP_000087.1 3 715
NR_046371.1 3 715
XM_006713499.2 XP_006713562.1 3 720
XM_006713500.3 XP_006713563.1 3 720
XM_006713501.2 XP_006713564.1 3 720
XM_011512435.1 XP_011510737.1 3 720
XM_017005734.1 XP_016861223.1 3 720
XM_017005735.1 XP_016861224.1 3 720
GenBank mRNA AK291219.1 - 3 497
AK299272.1 - 3 501
BC070304.1 - 3 668
BC094709.1 - 3 499
BC142714.1 - 3 471 40148840
BC146663.1 - 3 471 40148048
BC146801.1 - 3 495
M13699.1 - 3 462

View Full Product Details