Gene Transcripts
- Gene Symbol
- Tctex1d1
- Entrez Gene ID
- 67344
- Gene Name
- Tctex1 domain containing 1
- Gene Aliases
- 1700055O19Rik
- Chromosome Location
- Chr. 4: 102978388 - 103005594 on Build GRCm38
- UniGene ID
- Mm.111346
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163767.1 |
NP_001157239.1
|
4, 5 |
551 |
|
NM_001163768.1 |
NP_001157240.1
|
3, 4 |
458 |
|
NM_026100.3 |
NP_080376.1
|
4, 5 |
583 |
|
GenBank mRNA |
AK006804.1 |
- |
4, 5 |
552 |
|
AK015315.1 |
- |
4, 5 |
586 |
|
BB634690.1 |
- |
3, 4 |
470 |
|
BC048635.1 |
- |
4, 5 |
523 |
|
BC099549.1 |
- |
4, 5 |
557 |
30919464 |
BC132398.1 |
- |
4, 5 |
501 |
40130675 |
BC132400.1 |
- |
4, 5 |
501 |
40130677 |
BC145187.1 |
- |
4, 5 |
501 |
9053738 |
View Full Product Details