Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_207677.3 | NP_997560.3 | 5 | 1463 | |
GenBank mRNA | AK010701.1 | - | 5 | 1275 | |
AK043908.1 | - | 1308 | |||
AK089096.1 | - | 3 | 834 | ||
BC037043.1 | - | 4 | 1089 | ||
BC092250.1 | - | 5 | 1463 | 5043318 |