Gene Transcripts

Gene Symbol
Ndufa8
Entrez Gene ID
68375
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8
Gene Aliases
0610033L03Rik, AW261656
Chromosome Location
Chr. 2: 36036334 - 36049292 on Build GRCm38
UniGene ID
Mm.19834
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026703.2 NP_080979.1 2, 3 273
GenBank mRNA AK002744.1 - 2, 3 273
AK088463.1 - 2, 3 319
BC012416.1 - 2, 3 271 3584066
BC024673.1 - 2, 3 270 4195454

View Full Product Details