Gene Transcripts

Gene Symbol
Tspan11
Entrez Gene ID
68498
Gene Name
tetraspanin 11
Gene Aliases
1110014F12Rik, AU067806
Chromosome Location
Chr. 6: 127887622 - 127953031 on Build GRCm38
UniGene ID
Mm.174368
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026743.3 NP_081019.1 6, 7 885
GenBank mRNA AK003691.1 - 6, 7 737
AK028808.1 - 6, 7 776
AK053788.1 - 6, 7 872
AK090022.1 - 6, 7 885
BC057639.1 - 6, 7 727 4218418

View Full Product Details