Gene Transcripts
- Gene Symbol
- Kbtbd3
- Entrez Gene ID
- 69149
- Gene Name
- kelch repeat and BTB (POZ) domain containing 3
- Gene Aliases
- 2200003A07Rik, Bklhd3
- Chromosome Location
- Chr. 9: 4309743 - 4331829 on Build GRCm38
- UniGene ID
- Mm.25946
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164574.1 |
NP_001158046.1
|
2, 3 |
445 |
|
NM_026962.3 |
NP_081238.2
|
3, 4 |
477 |
|
GenBank mRNA |
AK008627.1 |
- |
2, 3 |
335 |
|
AK034334.1 |
- |
3, 4 |
385 |
|
AK078744.1 |
- |
2, 3 |
357 |
|
AK083497.1 |
- |
3, 4 |
459 |
|
AK168345.1 |
- |
2, 3 |
291 |
|
BB264869.2 |
- |
2, 3 |
446 |
|
BC024938.1 |
- |
2, 3 |
341 |
|
BC027392.1 |
- |
3, 4 |
381 |
4954712 |
BE650264.1 |
- |
3, 4 |
478 |
|
View Full Product Details