Gene Transcripts

Gene Symbol
Kcnip1
Entrez Gene ID
70357
Gene Name
Kv channel-interacting protein 1
Gene Aliases
KCHIP1, Kchip1.2
Chromosome Location
Chr. 11: 33629339 - 33993193 on Build GRCm38
UniGene ID
Mm.252514
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190885.1 NP_001177814.1 3 508
NM_001190886.1 NP_001177815.1 2 464
NM_001290690.1 NP_001277619.1 2 663
NM_027398.3 NP_081674.2 2 475
GenBank mRNA AB075041.1 - 2 105
AJ278534.1 - 1 33
AK014313.1 - 2 464
AK045948.1 - 3 509
AK046398.1 - 3 497
AK048178.1 - 2 424
AK081845.1 - 2 465
AK082267.1 - 2 260
AY050525.1 - 3 113
AY050526.1 - 2 80
AY171234.1 - 2 539
AY647242.1 - 2 307
BC034241.1 - 2 417 4503251
BU702946.1 - 1 161
DQ148493.1 - 2 417
DQ148494.1 - 2 465
DQ148495.1 - 3 113

View Full Product Details