Gene Transcripts

Gene Symbol
Mtfr2
Entrez Gene ID
71804
Gene Name
mitochondrial fission regulator 2
Gene Aliases
2610016C23Rik, 4933412C16Rik, Dufd1, Fam54a
Chromosome Location
Chr. 10: 20346989 - 20361669 on Build GRCm38
UniGene ID
Mm.262151
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027930.3 NP_082206.2 2, 3 214
GenBank mRNA AK011410.1 - 2, 3 209
AK016786.1 - 2, 3 259
BC019132.1 - 2, 3 218 5036908

View Full Product Details