Gene Transcripts

Gene Symbol
Mad2l2
Entrez Gene ID
71890
Gene Name
MAD2 mitotic arrest deficient-like 2
Gene Aliases
2310033C13Rik, G1-453-4, MAD2B, REV7, repro22
Chromosome Location
Chr. 4: 148130255 - 148145699 on Build GRCm38
UniGene ID
Mm.9648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK009587.1 - 4 560
AK150216.1 - 4 553
AK165627.1 - 4 364
BC011282.1 - 4 490 3484522
BC071264.1 - 6 490 6516319

View Full Product Details