Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028012.4 | NP_082288.1 | 5 | 649 | |
GenBank mRNA | AB055154.1 | - | 5 | 557 | |
AK009951.1 | - | 5 | 566 | ||
AK053612.1 | - | 565 | |||
AK088281.1 | - | 5 | 565 | ||
BC025538.1 | - | 5 | 593 | 5257949 |