Gene Transcripts

Gene Symbol
Rab4b
Entrez Gene ID
19342
Gene Name
RAB4B, member RAS oncogene family
Gene Aliases
1500031G17Rik
Chromosome Location
Chr. 7: 27168424 - 27178901 on Build GRCm38
UniGene ID
Mm.482114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029391.2 NP_083667.1 3 254
GenBank mRNA AB232592.1 - 3 113
AF408432.1 - 3 226
AK005314.1 - 3 254
AK140513.1 - 2 451
AK170552.1 - 3 268
BC007147.1 - 3 198 3600193
BC096654.1 - 3 237 5056766

View Full Product Details