Gene Transcripts

Gene Symbol
Fam204a
Entrez Gene ID
76539
Gene Name
family with sequence similarity 204, member A
Gene Aliases
2310065H12Rik, 2610015K05Rik, D19Ertd737e
Chromosome Location
Chr. 19: 60198586 - 60226726 on Build GRCm38
UniGene ID
Mm.290183
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029648.6 NP_083924.2 8 1281
GenBank mRNA AK011404.2 - 7 1171
AK075914.1 - 8 1282
BC061459.1 - 1080 6829614

View Full Product Details