Gene Transcripts

Gene Symbol
Stambpl1
Entrez Gene ID
76630
Gene Name
STAM binding protein like 1
Gene Aliases
1700095N21Rik, 8230401J17Rik, ALMalpha, AMSH-FP
Chromosome Location
Chr. 19: 34192190 - 34240657 on Build GRCm38
UniGene ID
Mm.130952
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029682.4 NP_083958.3 10 1614
GenBank mRNA AB010121.1 - 10 1309
AB010122.1 - 10 1273
AB066211.1 - 10 1614
AB066212.1 - 6 963
AB066213.1 - 10 1297
AK007087.1 - 6 947
BC018343.1 - 10 1340 4010423
BC023097.1 - 10 1583

View Full Product Details