Gene Transcripts

Gene Symbol
HLCS
Entrez Gene ID
3141
Gene Name
holocarboxylase synthetase
Gene Aliases
HCS
Chromosome Location
Chr. 21: 36750888 - 36990254 on Build GRCh38
UniGene ID
Hs.371350
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000411.6 NP_000402.3 5 569
NM_001242784.1 NP_001229713.1 5 1503
NM_001242785.1 NP_001229714.1 5 662
XM_005260953.3 XP_005261010.1 4 531
XM_005260955.3 XP_005261012.1 4 402
XM_005260956.3 XP_005261013.1 4 578
XM_006723994.2 XP_006724057.1 4 516
XM_006723995.1 XP_006724058.1 5 694
XM_011529538.1 XP_011527840.1 5 549
XM_011529539.2 XP_011527841.1 5 554
XM_011529540.2 XP_011527842.1 4 531
XM_011529541.2 XP_011527843.1 4 405
XM_017028330.1 XP_016883819.1 4 352
GenBank mRNA AJ001864.1 - 4 592
AK307940.1 - 4 530
AK314189.1 - 4 436
BC060787.1 - 5 527 30343493
D23672.2 - 5 560
D87328.1 - 5 1320
X80160.1 - 4 420

View Full Product Details