Gene Transcripts

Gene Symbol
HRG
Entrez Gene ID
3273
Gene Name
histidine rich glycoprotein
Gene Aliases
HPRG, HRGP, THPH11
Chromosome Location
Chr. 3: 186665952 - 186678240 on Build GRCh38
UniGene ID
Hs.1498
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000412.4 NP_000403.1 2 336
XM_005247415.3 XP_005247472.1 2 330
GenBank mRNA AK313381.1 - 2 306
BC069574.1 - 2 279 7262290
BC150591.1 - 2 277 9056961
CR749302.1 - 1 30
DA637240.1 - 2 330
DB128993.1 - 2 336
M13149.1 - 2 377

View Full Product Details