Gene Transcripts

Gene Symbol
Gpr63
Entrez Gene ID
81006
Gene Name
G protein-coupled receptor 63
Gene Aliases
PSP24beta, Psp24-2
Chromosome Location
Chr. 4: 24966541 - 25009233 on Build GRCm38
UniGene ID
Mm.160412
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030733.3 NP_109658.2 2 1160
GenBank mRNA AK036257.1 - 2 1161

View Full Product Details