Gene Transcripts

Gene Symbol
Plekha3
Entrez Gene ID
83435
Gene Name
pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 3
Gene Aliases
FAPP1
Chromosome Location
Chr. 2: 76675310 - 76697335 on Build GRCm38
UniGene ID
Mm.41636
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031256.3 NP_112546.1 8 1081
GenBank mRNA AF286163.1 - 8 870
BC012283.1 - 7 1340
BC031110.1 - 9 889 4981486

View Full Product Details