Gene Transcripts

Gene Symbol
Emc9
Entrez Gene ID
85308
Gene Name
ER membrane protein complex subunit 9
Gene Aliases
1500005A01Rik, Cgi112, Fam158a
Chromosome Location
Chr. 14: 55581519 - 55585356 on Build GRCm38
UniGene ID
Mm.40667
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033146.1 NP_149158.1 3, 4 482
GenBank mRNA AB054001.1 - 3, 4 528
AK005154.1 - 3, 4 482
AK144394.1 - 3, 4 510
AK154475.1 - 3, 4 531
BC024704.1 - 3, 4 458 4195108

View Full Product Details